Learn More
Rogo Sampaic™ Glass Volumetric Pipette with 0.1mL Graduation
Product Code. p-8008545
Click to view available options
Volume (Metric):
1 mL
5 mL
Unit Size:
Each
Product Code. 11782674
Brand: Rogo Sampaic™ DGG003
Specifications
Accuracy 0.05 mL
Calibrated Yes
Calibration 0.1mL
Class B
Graduated Yes
Graduation Color Brown
Graduations 0.1 mL
Color Blue
Volume (Metric) 5 mL
Length (Metric) 370 mm