Learn More
Rogo Sampaic™ Glass Volumetric Pipette with 0.1mL Graduation
Click to view available options
Volume (Metric):
1 mL
5 mL
Unit Size:
Each
Specifications
Specifications
| Accuracy | 0.05 mL |
| Calibrated | Yes |
| Calibration | 0.1mL |
| Class | B |
| Graduated | Yes |
| Graduation Color | Brown |
| Graduations | 0.1 mL |
| Color | Blue |
| Volume (Metric) | 5 mL |
| Length (Metric) | 370 mm |