Learn More
Hirschmann™ Class A Volumetric Flasks with Rim
Product Code. p-8011849
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Unit Size:
Each
Pack of 2
Product Code. 10767081
Brand: Hirschmann™ 2800198
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Rim are manufactured from durable DURAN™ glass tubing.
  • With blue graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Capacity (Metric) 10 L
Material Borosilicate Glass, Duran Glass
Height (Metric) 570 mm
Limit of Error ±2 mL
Tolerance ±2 mL
Graduated Yes
Class A