Learn More
Hirschmann™ Pasteur Pipettes with Cotton Plug
Product Code. 10673962
Change view
Click to view available options
Length (Metric):
150 mm
230 mm
Length (Metric) Tip:
40 mm
100 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Length (Metric) Tip Unit Size
10673962 230 mm 100 mm Pack of 1000
10450283 150 mm 40 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10673962 Supplier Hirschmann™ Supplier No. 9260201
  • ONLINE EXCLUSIVE PRICE
    155.00€
    106.00€ / Pack of 1000
    Save 49.00€
    31% Off
In Stock
Ideal for transferring samples to sample cups and for drop counting. Hirschmann™ Pasteur Pipettes are available in either long or short form.
  • Acc. to ISO 7712
  • With cotton plug
  • Long or Short form
  • With extended fine tip
Specifications
Aspirator Plug Cotton
Certifications/Compliance ISO 7712
Material Glass
Includes Finely tapered end and a cotton plug end
Length (Metric) Tip 100 mm
Quantity 1000 Pack
Top Style Fine drawn
Volume (Metric) 2 mL
Length (Metric) 230 mm
Type Pasteur Pipet