Learn More
Rogo Sampaic™ Glass Slides
Product Code. 11842340
Click to view available options
Color:
Blue
Green
Orange
Yellow
Material:
Glass
Unit Size:
Pack of 50
Product Code. 11842340
Brand: Rogo Sampaic™ BPB022
Only null left
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
Edges Cut
Material Glass
Color Orange
Quantity 50 Pack
Dimensions (L x W) 76 x 26 mm