Learn More
Gravis Glass Sample Bottles
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
12.5 mL
20 mL
25 mL
67 mL
96 mL
missing translation for 'unitSize'
Pack of 131
Pack of 140
Pack of 210
Pack of 86
Description
- Available in a range of capacities
Specifications
Specifications
| Color | Clear |
| Material | Glass |
| Volume (Metric) | 12.5 mL |
| No. per Pack | 20 |
| Quantity | 20 Pk. |