Learn More
Karl Hecht™ Glass Powder Funnel
Click to view available options
Diameter (Metric)Top:
5cm
8cm
12cm
Unit Size:
Each
Description
- Short, wide stem.
- AR™ soda glass construction.
Specifications
Specifications
| Diameter (Metric)Top | 5cm |
| Material | AR-GLAS |
| For Use With (Application) | Powdered materials |
| Product Type | Funnel |