Learn More
Cole-Parmer™ Glass MicroPipet
Product Code. 11878601 Shop All Cole-Parmer Products
Click to view available options
Volume (Metric):
1 μL
25 μL
50 μL
200 μL
Unit Size:
Pack of 100
Product Code. 11878601
Brand: Cole-Parmer™ 0795010
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Accuracy ±1%
Calibrated Yes
Diameter (Metric) Inner 0.19 mm
Diameter (Metric) Outer 0.635 mm
Material Glass
Quantity 100/Pk.
Volume (Metric) 1 μL
Length (Metric) 31.75 mm
Type Micropipet