Learn More
Rogo Sampaic™ Glass Lames
Product Code. 11832340
Click to view available options
Edges:
45°
90°, Cut
Unit Size:
Pack of 50
Product Code. 11832340
Brand: Rogo Sampaic™ BPB021
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
Edges 45°
Material Glass
Color White
Quantity 50 Pack
Dimensions (L x W) 76 x 26 mm