Learn More
Rogo Sampaic™ Glass Lames
Product Code. 11802350
Click to view available options
Edges:
45°
90°, Cut
Unit Size:
Pack of 50
Product Code. 11802350
Brand: Rogo Sampaic™ BPB027
Only null left
Add to basket
Edge
Add to basket
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
Edges 90°, Cut
Material Glass
Color White
Quantity 50 Pack
Dimensions (L x W) 76 x 26 mm