Learn More
Alla France™ Glass Hydrometer
Product Code. 11773183
Click to view available options
Specific Gravity Range:
0.800 to 0.900 g/mL
1.000 to 1.100 g/mL
1.100 to 1.200 g/mL
1.200 to 1.300 g/mL
Unit Size:
Each
Product Code. 11773183
Brand: Alla France™ 3010FG080/20qp
Use these hydrometers for a variety of applications. Alla France™ Glass Hydrometers feature a benchmark engraved on the stem to show any displacement of the scale.
  • General Purpose Hydrometers
  • 100 g/mL series
  • Density scale (g/mL)
  • Reading below meniscus
  • Without Certificate
Specifications
Quantity 1Each
Type Density Hydrometer
Accuracy ±0.002g/mL
Length (Metric) 300 mm
For Use With (Application) General use
Specific Gravity Range 0.800 to 0.900 g/mL