Learn More
Alla France™ Glass Hydrometer
Click to view available options
Specific Gravity Range:
0.800 to 0.900 g/mL
1.000 to 1.100 g/mL
1.100 to 1.200 g/mL
1.200 to 1.300 g/mL
Unit Size:
Each
Description
- General Purpose Hydrometers
- 100 g/mL series
- Density scale (g/mL)
- Reading below meniscus
- Without Certificate
Specifications
Specifications
| Quantity | 1Each |
| Type | Density Hydrometer |
| Accuracy | ±0.002g/mL |
| Length (Metric) | 300 mm |
| For Use With (Application) | General use |
| Specific Gravity Range | 1.100 to 1.200 g/mL |