Specifications
| Capacity (Metric) | 1000 mL |
| Material | Glass |
| Certifications/Compliance | USP Type I and ASTM E 438 Type I, Class A |
| For Use With (Application) | 90mm Vacuum filter holder assemblies |
| Product Type | Funnel |