Learn More
Macherey-Nagel™ Glass Flash Chromatography Column
Click to view available options
Length (Metric):
200 mm
300 mm
400 mm
450 mm
Style:
I.D.: 20 mm
I.D.: 25 mm
I.D.: 30 mm
I.D.: 40 mm
Unit Size:
Each
Specifications
Specifications
| Length (Metric) | 200 mm |
| Material | Glass |
| Style | I.D.: 25 mm |
| Type | Flash Column |
| Includes | with Adapter and PTFE Tap |
| Column Type | Flash Chromatography Glass Column |
| For Use With (Application) | Suited The Separation of Compounds up to Gram Levels No Expensive Equipment Required |