Learn More
Macherey-Nagel™ Glass Flash Chromatography Column
Product Code. 10164801
Click to view available options
Length (Metric):
200 mm
300 mm
400 mm
450 mm
Style:
I.D.: 20 mm
I.D.: 25 mm
I.D.: 30 mm
I.D.: 40 mm
Unit Size:
Each
Product Code. 10164801
Brand: Macherey-Nagel™ 727404
Specifications
Length (Metric) 300 mm
Material Glass
Style I.D.: 30 mm
Type Flash Column
Includes with Adapter and PTFE Tap
Column Type Flash Chromatography Glass Column
For Use With (Application) Suited The Separation of Compounds up to Gram Levels No Expensive Equipment Required