Learn More
Fisherbrand™ Glass-Encased Stir Bar
Product Code. 11828902 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
6 mm
12 mm
19 mm
25 mm
45 mm
60 mm
Diameter (Metric):
5 mm
6 mm
8 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 11828902
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use with very abrasive media that may erode PTFE. The glass-encased magnetic stir bar is cylindrical for a wide variety of uses.
  • Stable, general-purpose magnetic stir bar
  • Use with very abrasive media that may erode PTFE
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color Clear
Description Glass encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 5 mm
Length (Metric) 12 mm
Quantity 5/Pk.
Shape Cylinder