Learn More
3M™ Glass Cleaner and Protector
Product Code. 17217640 Shop All 3M Products
Change view
Click to view available options
Volume (Metric):
950 mL
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17217640 950 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17217640 Supplier 3M™ Supplier No. 7100038228
  • ONLINE EXCLUSIVE PRICE
    183.00€
    111.61€ / Pack of 12
    Save 71.39€
    39% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Glass Cleaner and Protector
  • Cleans and protects glass for a faster, longer lasting shine
  • Makes light work of fingerprints, lipstick and even permanent marker
  • Reduces fogging on mirrors and other glass surfaces
  • Dries faster than other glass cleaners tested, saving time
  • Non-ammoniated formula with a pleasant smell
  • The first commercial glass cleaner with Scotchgard Protection
  • Dries faster than other glass cleaners tested, saving time
TRUSTED_SUSTAINABILITY
Specifications
Description Ready -To-Use
Volume (Metric) 950 mL