Learn More
Hecht Karl™ Glass Class AS Volumetric Pipet
Brand:  Hecht Karl™ 42556032
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10033811
  • 5.14€ / Each
Estimated Shipment: 17-09-2024
Specifications
Yes
AS
Yes
1 Pack
5 mL
DIN EN ISO 648, Conformity certified
Glass
Amber
0.015 mL
Volumetric Pipet