Learn More
Marienfeld Superior™ Glass Capillary Tubes for the Determination of Melting Point
Click to view available options
Length (Metric) Tube:
70 mm
75 mm
80 mm
Diameter (Metric) Inner:
0.42 mm
0.9 mm
1 mm
1.2 mm
1.5 mm
Product Type:
Capillary Tubes (Both Ends Open)
Capillaty Tubes (One End Closed)
Unit Size:
Pack of 1000
Description
- With rectangular cut edges
- For single use
- 100 capillaries in a dispenser vial, 10 vials in a box
Specifications
Specifications
| Material | Glass |
| Product Type | Capillaty Tubes (One End Closed) |
| Diameter (Metric) Inner | 1 mm |
| For Use With (Application) | For Determination Of Melting Point |
| Length (Metric) Tube | 80 mm |
| Quantity | 1000 |