Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper
Product Code. 10611923
Click to view available options
Capacity (Metric):
10 mL
10,000 mL
100 mL
1000 mL
20 mL
200 mL
2000 mL
25 mL
250 mL
5 mL
50 mL
500 mL
5000 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
19/32 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
NS 14/23
Unit Size:
Each
Pack of 2
Product Code. 10611923
Brand: Hirschmann™ 2822198
Achieve precise standard solutions. Hirschmann™ Class A Volumetric Flasks with Poly Stopper is standardized to contain or to deliver a definite amount of liquid. Flask has silver diffusion ink with brown graduations.
  • With polyethylene stopper
  • With amber stain graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Calibration To Contain
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Color Permanent amber markings with silver diffusion color
Capacity (Metric) 10,000 mL
Material Borosilicate Glass (3.3)
Height (Metric) 570 ± 5 mm
Includes Polyethylene Stopper
Limit of Error ± 2 mL
No. per Pack 1/Pack
Standard Taper Joint 45/40 NS Standard Ground
Tolerance ± 2 mL
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Sterility Non-sterile
Class A
Bottom Shape Round