Learn More
RETSCH Glass Beads for Mills
Product Code. 11726175 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
0.10 to 0.25 mm
0.25 to 0.50 mm
0.50 to 0.75 mm
0.75 to 1 mm
1 to 1.5 mm
Unit Size:
500g
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11726175 0.10 to 0.25 mm Each
11736175 0.25 to 0.50 mm Each
11756175 0.75 to 1 mm 500g
10782701 1 to 1.5 mm 500g
11746175 0.50 to 0.75 mm 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11726175 Supplier RETSCH Supplier No. 22.222.0001
  • ONLINE EXCLUSIVE PRICE
    64.90€
    60.83€ / Pack of 1
    Save 4.07€
    6% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
For use with Retsch MM 200 Mixer Mills.
  • Ideal for disruption of biological cells
Specifications
Type Beads
Material Glass
Diameter (Metric) 0.10 to 0.25 mm
For Use With (Equipment) Mixer Mill MM 200
Quantity 500 g