Learn More
Edmund Bühler GmbH™ Glass Beads
Product Code. 10526332
Change view
Click to view available options
Diameter (Metric):
0.1 mm
0.30 mm
0.50 mm
Unit Size:
1kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10526332 0.1 mm 1kg
10115751 0.50 mm 1kg
10648841 0.30 mm 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10526332 Supplier Edmund Bühler GmbH™ Supplier No. 0001 063
Only null left
Add to basket
Add to basket
For the dissolution of samples into powder
  • Glass construction
  • PbO-free
  • 1kg each in plastic bottle
  • For dissolution of samples into powder
Specifications
Material Glass
Diameter (Metric) 0.1 mm
Description PbO-free