missing translation for 'onlineSavingsMsg'
Learn More

Gimeracil, TRC

Product Code. 30403549
Change view
Click to view available options
Quantity:
5 mg
50 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403549 5 mg
30404805 50 mg
30391785 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403549 Supplier Toronto Research Chemicals Supplier No. TRCG3780005MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103766-25-2
Molecular Formula C5 H4 Cl N O2
Molecular Weight (g/mol) 145.54
IUPAC Name 5-chloro-4-hydroxy-1H-pyridin-2-one
SMILES OC1=CC(=O)NC=C1Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H4ClNO2/c6-3-2-7-5(9)1-4(3)8/h1-2H,(H2,7,8,9)
Formula Weight 144.9931
Chemical Name or Material Gimeracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.