Learn More
Gilson™ PIPETMANTM TOWERPACKTM Refill System
GREENER_CHOICE
Product Code. 10113532 Shop All Gilson Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
100 to 1000 μL
2 to 200 μL
20 to 300 μL
For Use With (Equipment):
Universal Fit
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 672
Pack of 960
Product Code. 10113532
Brand: Gilson™ F167101
Edge
Differentiates tip models with color-coded racks and indicates the compatible Gilson pipet models. GilsonTM PIPETMANTM TOWERPACKTM Refill System is available in autoclavable and sterilized versions.
  • Color-coded racks to differentiate tip models and indicate the compatible GilsonTM pipet models
  • Available in autoclavable and sterilized versions
  • Fits Universal Reload box (Cat. No. F167100G, available separately)
  • Sterilized Tower Pack is individually wrapped in medical device-quality plastic to help protect tips from contamination
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.1 to 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 4.21 in.
Length (Metric) 10.7 cm
Diameter (Metric) Outer .4 mm
Format Blister Refill
Tip Style Standard
Quantity 960 (10x96)
Product Type Tower Pack™ Pipet Refill System
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes