Learn More
Gilson™ PIPETMANTM L Multichannel Pipet
Shop All Gilson Products
Click to view available options
Color:
Green
Red
Yellow
Number of Channels:
8
12
Volume (Metric):
0.5 to 10 μL
2 to 20 μL
20 to 200 μL
20 to 300 μL
Unit Size:
Each
Description
- Gilson–patented volume locking system guarantees no accidental change in volume while pipetting.
- Adjustable tip ejector provides comfort for right- and left-handed users.
- Redesigned handle fits naturally in your hand, no matter the size.
- Unique internal design applies even, equal pressure from either side of the pipette, rather than on the center of the channels like other multichannels do.
- 2D identification code tracks product reference number, date of manufacture, serial number and maximum volume of pipet.
- Available in 8–and 12–channel covering a volume range from 0.5 to 300μL.
Specifications
Specifications
| Compatible Tips | PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST |
| Adjustable Tip Spacing | No |
| Autoclavable | Autoclavable |
| Color | Green |
| Dispensing Speed | Manual |
| Disposable | Reusable |
| For Use With | Pipette Tip 50 to 1000 μL |
| Grip Type | Ergonomic |
| Material | Polypropylene |
| Model | P12x300L |
| Show More |