Learn More
Gilson™ PIPETMANTM EasyPackTM Tips
GREENER_CHOICE
Product Code. 10614353 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
1 to 10 mL
1 to 5 mL
100 to 1000 μL
100 to 1200 μL
2 to 200 μL
20 to 300 μL
Format:
Bagged
Quantity:
1000 (5x200)
200
200 (4x50)
334
Unit Size:
Pack of 1000
Pack of 1002
Pack of 200
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Format Quantity Unit Size
10614353 0.1 to 10 μL Bagged 1000 (5x200) Pack of 1000
10367132 1 to 10 mL Bagged 200 (4x50) Pack of 200
12629591 100 to 1200 μL Bagged 200 Pack of 1000
10521103 0.1 to 20 μL Bagged 200 Pack of 1000
10357852 1 to 5 mL Bagged 334 Pack of 1002
10001962 100 to 1000 μL Bagged 1000 (5x200) Pack of 1000
10367802 2 to 200 μL Bagged 1000 (5x200) Pack of 1000
10245832 20 to 300 μL Bagged 1000 (5x200) Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10614353 Supplier Gilson™ Supplier No. F161631
  • ONLINE EXCLUSIVE PRICE
    56.24€
    42.00€ / Pack of 1000
    Save 14.24€
    25% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Ensure optimal performance of PIPETMAN pipettes across the full volume range with PIPETMANTM EasyPackTM Tips.
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips).
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.1 to 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 3.74 in.
Length (Metric) 9.5 cm
Diameter (Metric) Outer .4 mm
Format Bagged
Tip Style Universal
Packaging 5 Bags of 200 Tips
Quantity 1000 (5x200)
Product Type Pipet Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Gilson Pipetman
Graduated Yes
Product Line Pipetman EasyPack
Material Polypropylene
Graduations 2 μL, 10 μL