Learn More
Gilson™ PIPETMANTM EasyPackTM Tips
GREENER_CHOICE
Product Code. 10521103 Shop All Gilson Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
1 to 10 mL
1 to 5 mL
100 to 1000 μL
100 to 1200 μL
2 to 200 μL
20 to 300 μL
Format:
Bagged
Quantity:
1000 (5x200)
200
200 (4x50)
334
Unit Size:
Pack of 1000
Pack of 1002
Pack of 200
Product Code. 10521103
Brand: Gilson™ F161451
Ensure optimal performance of PIPETMAN pipettes across the full volume range with PIPETMANTM EasyPackTM Tips.
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips).
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.1 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model DL10 EASYPACK
Length (English) 4.72 in.
Length (Metric) 12 cm
Diameter (Metric) Outer .04 mm
Format Bagged
Tip Style Universal
Packaging 5 Bags of 200 Tips
Quantity 200
Product Type Pipet Tips
Color Code Red
Disposable Reusable
For Use With (Equipment) Gilson Pipetman
Graduated Yes
Product Line Pipetman EcoPack
Material Polypropylene
Graduations 2 μL, 10 μL