Specifications
| Material | Vinyl |
| Sterility | Sterile |
| No. per Pack | 50 |
| Dimensions (L x W) | 79.4 x 145.5 mm |
| Product Type | Microplate Sealing Film |
| Thickness | 100 μm |
| Pierceable | Yes |
| Temperature Range (Metric) | -40°C to +90°C |