Learn More
Masterflex™ Gilmont™ Nylon Male Luer Fitting
Product Code. 12605750 Shop All Masterflex Products
Change view
Click to view available options
For Use With:
Male luer x 1/16 in. Hose Barb
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
12605750 Male luer x 1/16 in. Hose Barb Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12605750 Supplier Masterflex™ Supplier No. 4550522
  • ONLINE EXCLUSIVE PRICE
    19.05€
    18.26€ / Pack of 25
    Save 0.79€
    4% Off
Estimated Shipment: 19-02-2027
Specifications
Material Nylon
For Use With Male luer x 1/16 in. Hose Barb
Packaging Quantity 25 Pack
Product Type Male Luer Fitting