Learn More
Gilac™ Gilactiv™ Bedpan
Product Code. 17938881
Change view
Click to view available options
Volume (Metric):
2.5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17938881 2.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17938881 Supplier Gilac™ Supplier No. G101190
  • ONLINE EXCLUSIVE PRICE
    16.67€
    10.98€ / Pack of 1
    Save 5.69€
    34% Off
Estimated Shipment: 09-12-2026
Gilactiv™ Bedpan is treated with the exclusive Sanitized™ technology, which protects the bedpan's surface against viruses and bacteria.
  • Suitable for hospital use.
  • Translucent, level visible, with volume graduations.
  • Rounded design and very thick.
  • Not easy to break when dropped.
  • Outstanding durability even in the intensive use cycle.
  • Product is disinfected in a bedpan washer.
  • Researched product ergonomics ensure optimum cleaning.
  • Equipment at room temperature to avoid the cold sensation experienced with stainless steel equivalents.
Specifications
Product Type Bedpan
Color Blue
Material Polyethylene
Sterility Non-sterile
Volume (English) 0.7 gal.
Volume (Metric) 2.5 L