Learn More
Gilac™ Gilactiv™ Bedpan Lid
Product Code. 17928641
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17928641 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17928641 Supplier Gilac™ Supplier No. G101390
  • ONLINE EXCLUSIVE PRICE
    8.17€
    5.37€ / Pack of 1
    Save 2.80€
    34% Off
Estimated Shipment: 09-12-2026
Gilactiv™ Bedpan Lid is treated with the exclusive Sanitized™ technology, which protects the lid's surface against viruses and bacteria.
  • Suitable for hospital use.
  • Translucent, level visible, with volume graduations.
  • Rounded design and very thick.
  • Not easy to break when dropped.
  • Outstanding durability even in the intensive use cycle.
  • Product is disinfected in a bedpan washer.
  • Researched product ergonomics ensure optimum cleaning.
  • Equipment at room temperature to avoid the cold sensation experienced with stainless steel equivalents.
Specifications
Product Type Bedpan Lid
Color Blue
Material Polyethylene
Sterility Non-sterile
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.