Learn More
Brady™ GHS Symbol - Health Hazard, Individual Markers on a Roll
Product Code. 15462044
Click to view available options
Material:
Polyester
Unit Size:
1 roll
Product Code. 15462044
Brand: Brady™ 834182
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Pre-printed labels to identify your chemical product containers.
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Alternate Part Number PIC 1805-21*21-B7541-RLL-DC
Finish Gloss
Material Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 21 mm
Thickness 0.099 mm
Width (Metric) 21 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Color Black on White