Learn More
Vilber Lourmat™ Germicidal UV Lamp
Click to view available options
Length (Metric):
500 mm
505 mm
980 mm
1290 mm
Wattage:
15 W
30 W
45 W
60 W
80 W
120 W
Unit Size:
Each
Specifications
Specifications
| Voltage | 230 V |
| Wattage | 30 W |
| Wavelength | 254 nm |
| Depth (Metric) | 140 mm |
| Electrical Requirements | 30 W, 230 V, 50 Hz |
| Height (Metric) | 100 mm |
| Hertz | 50 Hz |
| Length (Metric) | 980 mm |
| Lighting Intensity | 60 mW/cm2 |
| Packaging Quantity | 1 Ea. |
| Show More |