Learn More
Thermo Scientific™ Gentamicin Sulfate, Ultrapure
Inhibits normal protein synthesis
Brand: Thermo Scientific™ J16051.06
Additional Details : CAS : 1405-41-0 Weight : 1.00000kg
Description
- Bactericidal
- Stable
- Autoclavable
- Effective when combined with antibiotic that interferes with cell wall synthesis
Specifications
Requires no refrigeration | |
≥590 μg/mg (Dry basis) | |
Antibiotic | |
3.5 to 5.5 | |
5 g |
Bactericidal | |
Powder | |
Plastic bottle | |
Ultrapure |