Learn More
Genomic DNA isolation, Lysis Buffer C1, Macherey-Nagel™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Chemical Name or Material | Lysis Buffer C1 |
| For Use With (Application) | Genomic DNA Isolation |
| Quantity | 100 mL |