Learn More
3M™ General Purpose Vinyl Tape 764i
Product Code. 17257780
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Unit Size:
24 rolls
Product Code. 17257780
Brand: 3M™ 7000144707
Used for all non-critical applications, including colour coding, bundling and safety marking. Our non-corrosive, flexible vinyl tape is ideal for protecting parts, equipment and products from abrasions.
  • Use for taping in all non-critical applications
  • Protects parts, equipment and products from abrasions
  • Bundle items, bundle and temporarily mark parts, machines, plus much more
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas. Lots of different colours available such as yellow, red, blue, green, black, white, etc
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 50 mm x 33 m
Length (Metric) 33 m
Thickness 0.13 mm
Width (Metric) 50 mm
Color White