Description
- Sterile tubes have attached blue screwcap
- Black printed graduations in 0.5mL increments
- White printed writing space
- Autoclavable (without caps)
- Spin Rate: 8500 RCF
- Temperature Range: -40°C to +80°C
Specifications
Specifications
| Closure Color | Blue |
| Capacity (Metric) | 15 mL |
| Material | Polypropylene |
| Packaging | Racked |
| Quantity | 500/Cs. |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| Length (Metric) | 120 mm |
| Bottom Shape | Conical |
| Closure Type | Screw Cap |
| Show More |