Learn More
Thermo Scientific™ General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
2835.00€ - 6170.00€
Specifications
Material | See individual centrifuge, rotor and accessory data |
---|---|
Net Weight | See individual components of package |
For Use With (Equipment) | Purchase of Thermo Scientific General Purpose Centrifuge required for operation |
Warranty | See Manual |
Biocontainment | Yes |
Product Code | Brand | Description | Type | Capacity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Type | Capacity | Price | Quantity & Availability | |||
16350778
|
Thermo Scientific™
75016103 |
Thermo Scientific TX-1000 Blood Tube Package | TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters | Max in TX-1000: 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
6170.00€
1 unit |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
16360778
|
Thermo Scientific™
75016104 |
Thermo Scientific TX-1000 Microplate Package | TX-1000 with Buckets, ClickSeal Biocontainment lids, microplate Adapters | Max in TX-1000: 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
5370.00€
1 unit |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
16370778
|
Thermo Scientific™
75016105 |
Thermo Scientific TX-1000 Cell Culture Package | TX-1000 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adapters | Max in TX-1000: 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
6120.00€
1 unit |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
16380778
|
Thermo Scientific™
75016106 |
Thermo Scientific TX-200 Cell Culture Package | TX-200 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adapters | Max in TX-200: 20 x 15 mL Conical Tubes, 4 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
2835.00€
1 unit |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
16390778
|
Thermo Scientific™
75016107 |
Thermo Scientific TX-400 Blood Tube Package | TX-400 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters | Max in TX-400: 76 x 5/7 mL Blood Tubes, 56 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
5100.00€
1 unit |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
16300788
|
Thermo Scientific™
75016108 |
Thermo Scientific TX-400 Cell Culture Package | TX-400 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adapters | Max in TX-400: 36 x 15 mL Conical Tubes, 16 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
5100.00€
1 unit |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
16310788
|
Thermo Scientific™
75016109 |
Thermo Scientific TX-750 Blood Tube Package | TX-750 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters | Max in TX-750: 148 x 5/7 mL Blood Tubes, 84 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
5505.00€
1 unit |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
16320788
|
Thermo Scientific™
75016110 |
Thermo Scientific TX-750 Cell Culture Package | TX-750 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adapters | Max in TX-750: 56 x 15 mL Conical Tubes, 28 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
5505.00€
1 unit |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
16330788
|
Thermo Scientific™
75016111 |
Thermo Scientific TX-200 Blood Tube Package with TX-200 Swinging Bucket Rotor | TX-200 with 4 Buckets, 4 ClickSeal Biocontainment lids, 5/7 and 10 mL Blood Tube Adapters | Max in TX-200: 36 x 5/7 ml Blood Tubes, 28 x 10 mL Blood tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) |
2835.00€
1 unit |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
Description
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
See individual centrifuge, rotor and accessory data | |
Purchase of Thermo Scientific General Purpose Centrifuge required for operation | |
Yes |
See individual components of package | |
See Manual |