Learn More
Thermo Scientific™ General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
Brand: Thermo Scientific™ 75016111
2033.15 EUR valid until 2024-12-31
Use promo code "21988" to get your promotional price.
Additional Details : Weight : 4.00000kg
Description
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
Thermo Scientific TX-200 Blood Tube Package with TX-200 Swinging Bucket Rotor | |
TX-200 with 4 Buckets, 4 ClickSeal Biocontainment lids, 5/7 and 10 mL Blood Tube Adapters | |
See individual components of package | |
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | |
See Manual |
See individual centrifuge, rotor and accessory data | |
Max in TX-200: 36 x 5/7 ml Blood Tubes, 28 x 10 mL Blood tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) | |
Purchase of Thermo Scientific General Purpose Centrifuge required for operation | |
Package Includes: TX-200 rotor cross w/ Auto=Lock, 4 Buckets, 4 ClickSeal biocontainment lids, 5/7mL blood tube adapters (set/4, 36 tube capacity), 10mL blood tube adapters (set/4, 28 tube capacity) | |
Yes |