Learn More
Gemfibrozil 1-O-Beta-Glucuronide-d6, TRC
Product Code. 30240681
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.50mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240681 10 mg 10mg
30240872 0.5 mg 0.50mg
30240891 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240681 Supplier Toronto Research Chemicals Supplier No. TRCG30576210MG
 Restricted Item
Estimated Shipment: 14-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1703747-47-0
Molecular Formula C21H24D6O9
Molecular Weight (g/mol) 432.49
Synonym 1-[5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoate] β-D-Glucopyranuroic Acid-d6, Gemfibrozil-d6 Glucuronide;
SMILES CC1=C(OCCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C(O[C@@H]2O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]2O)=O)C=C(C)C=C1
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C21H30O9/c1-11-6-7-12(2)13(10-11)28-9-5-8-21(3,4)20(27)30-19-16(24)14(22)15(23)17(29-19)18(25)26/h6-7,10,14-17,19,22-24H,5,8-9H2,1-4H3,(H,25,26)/t14-,15-,16+,17-,19-/m0/s1/i3D3,4D3
Formula Weight 432.2266
Chemical Name or Material Gemfibrozil 1-O-beta-Glucuronide-d6