Learn More
Interscience™ GeckoGrip Set for DiluFlow
Product Code. 16091792
Change view
Click to view available options
For Use With (Equipment):
DiluFlow, DiluFlow Pro, DiluFlow Elite
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16091792 DiluFlow, DiluFlow Pro, DiluFlow Elite Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16091792 Supplier Interscience™ Supplier No. 505009
  • ONLINE EXCLUSIVE PRICE
    105.00€
    103.08€ / Pack of 10
    Save 1.92€
    1% Off
View all promotions
Estimated Shipment: 08-01-2027
Set of 10 GeckoGrip adhesives for use with the DiluFlow BagOpen 400 and 3500.
  • Ensure your BagOpen is stabilized with GeckoGrip adhesives
Specifications
Product Type Bag Adhesive
Autoclavable Not Autoclavable
For Use With (Application) Stabilizing BagOpen
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite
Quantity 10 Adhesives/Pk.