Learn More
Masterflex Gear Pump Head, Cavity Style, Stainless Steel, PTF
Brand:  Masterflex 74004-01
Gear Pump Head, Cavity Style, Stainless Steel, PTFE Gear, PTFE Seal, 0.64mL/rev
Product Code. 15633816
  • ONLINE EXCLUSIVE PRICE
    3355.00€
    2774.55€ / Each
    Save 580.45€
    17% Off
Only null left
Estimated Shipment: 18-03-2026