missing translation for 'onlineSavingsMsg'
Learn More

GDI2 Mouse anti-Human, Mouse, Clone: 4B8F3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60078-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16867045

  • 399.00€ / 150µL
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

This antibody can bind both GDIs for the close sequences.

Regulates the GDP/GTP exchange reaction of most Rab proteins by inhibiting the dissociation of GDP from them, and the subsequent binding of GTP to them.
Specifications

Specifications

GDI2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P50395, Q61598
Gdi2
GDI2 Fusion Protein Ag4613
150 μL
Primary
Human, Mouse
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
4B8F3
Unconjugated
GDI2
GDI 2, GDI2, GDP dissociation inhibitor 2, Rab GDI beta, RABGDIB
Mouse
Caprylic Acid Precipitation
RUO
14569, 2665
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers