Learn More
Trajan™ Sealing Ring for GC Ferrule
Product Code. 15298873
Change view
Click to view available options
Diameter (Metric) Inner:
1.59 mm
6.35 mm
Length (Metric):
1.7 mm
2.2 mm
Width (Metric):
3.2 mm
9.9 mm
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (Metric) Width (Metric) Unit Size
15298873 1.59 mm 1.7 mm 3.2 mm Pack of 10
10578102 6.35 mm 2.2 mm 9.9 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15298873 Supplier Trajan™ Supplier No. 0726520
 Restricted Item
Estimated Shipment: 22-12-2026
Graphite sealing ring for use with GC Ferrules
  • For use with GC Ferrules
  • 100% Graphite Sealing ring for 1/4 in. fittings
  • Supplied in packs of 10
Specifications
Diameter (English) Inner 1/16 in.
Diameter (Metric) Inner 1.59 mm
Length (Metric) 1.7 mm
Material 100% Graphite
Max. Temperature (Metric) 450°C
Product Type Sealing Ring
Width (Metric) 3.2 mm