Learn More
Trajan™ BP624 GC Capillary Columns
Product Code. 12925581
Click to view available options
Length (Metric):
30 m
30 mm
50 m
60 m
Diameter (Metric) Inner:
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
1.4 μm
1.8 μm
3 μm
1.2 mm
3 mm
Unit Size:
Each
Product Code. 12925581
Brand: Trajan™ 054840
SGE BP624 GC Capillary Columns - A polar phase designed for EPA volatile organics analysis
  • Polar phase
  • EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
  • Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
Max. Temperature 230/240°C
Diameter (Metric) Inner 0.25 mm
Film Thickness 1.4 μm
Length (Metric) 30 m