Learn More
GBR-12909 Dihydrochloride, TRC
Product Code. 17943795
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17943795 5 mg 5mg
17971215 50 mg 50mg
17912065 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17943795 Supplier Toronto Research Chemicals Supplier No. TRCG2680055MG
 Restricted Item
Estimated Shipment: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67469-78-7
Molecular Formula C28 H32 F2 N2 O . 2(H Cl)
Molecular Weight (g/mol) 523.48
Synonym GBR-12909 Dihydrochloride, 1-[2-[Bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)-piperazine Dihydrochloride (9CI), GBR 12909, I 893, Vanoxerine Dihydrochloride, 1-[2-[Bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)-piperazine Hydrochloride (1:2)
IUPAC Name 1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine;hydrochloride
SMILES Cl.Cl.Fc1ccc(cc1)C(OCCN2CCN(CCCc3ccccc3)CC2)c4ccc(F)cc4
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C28H32F2N2O.2ClH/c29-26-12-8-24(9-13-26)28(25-10-14-27(30)15-11-25)33-22-21-32-19-17-31(18-20-32)16-4-7-23-5-2-1-3-6-23;;/h1-3,5-6,8-15,28H,4,7,16-22H2;2*1H
Formula Weight 522.2016
Chemical Name or Material 1-[2-[Bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)-piperazine Dihydrochloride