Learn More
Thermo Scientific™ Gator A2 Large System 24-Slot Rapid Load UVT Gel Trays, with gasketed End Gates
Product Code. 10330124 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
A2 and A5 Large Gel Electrophoresis System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10330124 A2 and A5 Large Gel Electrophoresis System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10330124 Supplier Thermo Scientific™ Supplier No. A2RLUVT
  • ONLINE EXCLUSIVE PRICE
    302.00€
    269.00€ / Pack of 1
    Save 33.00€
    10% Off
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
A2 and A5 Large Gel 24-slot Rapid Load UVT Gel Trays enable multiple sample loading.
Thermo Scientific™Owl A2 and A5 Large Gel 24-slot Rapid Load UVT Gel Trays enable multiple sample loading.
Each side of UVT gel tray is fitted with a gasketed end gate which provides a leakproof seal without tape. Built in leveling ensures the casting of flat, uniformly thick gels.
TRUSTED_SUSTAINABILITY
Specifications
Description 24-slot rapid load UVT gel tray with gasketed end gates
For Use With (Equipment) A2 and A5 Large Gel Electrophoresis System
Type Gel Tray