Learn More
Karl Hecht Drechsel gas washing bottles 100ml
Brand:  Karl Hecht 41469012
Drechsel gas washing bottles 100ml
Product Code. 10023631
  • ONLINE EXCLUSIVE PRICE
    85.05€
    58.69€ / Pack of 1
    Save 26.36€
    30% Off
 Restricted Item
Estimated Shipment: 10-12-2026