Learn More
Thermo Scientific™ Cover for Precision™ Water Baths
Product Code. 15335897 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Gable Cover
Large Gassing Hood
Small Gassing Hood
Material:
Polycarbonate
Stainless Steel
For Use With (Equipment):
Precision 10L Water Baths
Precision 15L Shaking Water Baths
Precision 20L and 28L Water Baths
Precision 2L Water Bath
Precision 2L and 5L Water Baths
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material For Use With (Equipment) unitSize
15335897 Small Gassing Hood Stainless Steel Precision 15L Shaking Water Baths Each
15345897 Large Gassing Hood Stainless Steel Precision 15L Shaking Water Baths Each
15345887 Gable Cover Polycarbonate Precision 2L Water Bath Each
15355887 Gable Cover Polycarbonate Precision 2L and 5L Water Baths Each
15365887 Gable Cover Polycarbonate Precision 10L Water Baths Each
15375887 Gable Cover Polycarbonate Precision 20L and 28L Water Baths Each
15385887 Gable Cover Stainless Steel Precision 2L Water Bath Each
15325897 Gable Cover Stainless Steel Precision 20L and 28L Water Baths Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15335897 Supplier Thermo Scientific™ Supplier No. 3162639
Only null left
Add to basket
Add to basket
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Warranty
1 year
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Precision 15L Shaking Water Baths
Type Small Gassing Hood
Material Stainless Steel