Learn More
Afora™ Gas Washing Bottles
Product Code. 11922408
Change view
Click to view available options
Quantity:
1 Pack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11922408 1 Pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11922408 Supplier Afora™ Supplier No. 5223/3
  • ONLINE EXCLUSIVE PRICE
    105.12€
    94.46€ / Pack of 1
    Save 10.66€
    10% Off
Estimated Shipment: 23-12-2026
Specifications
Capacity (Metric) 250 mL
Type Gas Washing Bottle
Standard Taper Joint Female 29/32
Quantity 1 Pack